gRNA Design Details
- Target DNA Sequence
- CGGGTGAACCCCAAGTCTTA
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 24270057 - 24270079 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RABGGTA
- Entrez Gene ID
-
5875
- Gene Name
- Rab geranylgeranyltransferase alpha subunit
- Gene Aliases
-
PTAR3
- Chromosome Location
-
Chr.14: 24265538 - 24271627 on GRCh38
- UniGene ID
- Hs.377992
- Species
-
Human