gRNA Design Details
- Target DNA Sequence
- TGGCAAGGAGGGCACCTATT
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 44327961 - 44327983 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.26/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Dpagt1
- Entrez Gene ID
-
13478
- Gene Name
- dolichyl-phosphate (UDP-N-acetylglucosamine) acetylglucosaminephosphotransferase 1 (GlcNAc-1-P transferase)
- Gene Aliases
-
AU021132, Dpagt2, GPT, Gnpta
- Chromosome Location
-
Chr.9: 44326017 - 44333606 on GRCm38
- UniGene ID
- Mm.18353
- Species
-
Mouse