gRNA Design Details
- Target DNA Sequence
- GTCGGGTAGAGCAGATTCTG
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 179142940 - 179142962 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ABL2
- Entrez Gene ID
-
27
- Gene Name
- ABL proto-oncogene 2, non-receptor tyrosine kinase
- Gene Aliases
-
ABLL, ARG
- Chromosome Location
-
Chr.1: 179099327 - 179229693 on GRCh38
- UniGene ID
- Hs.159472
- Species
-
Human