Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_000662.7 | NP_000653.3 | 3 |
NM_001160170.3 | NP_001153642.1 | 5 | |
NM_001160171.3 | NP_001153643.1 | 4 | |
NM_001160172.3 | NP_001153644.1 | 5 | |
NM_001160173.3 | NP_001153645.1 | 4 | |
NM_001160174.2 | NP_001153646.1 | 1 | |
NM_001160175.3 | NP_001153647.1 | 5 | |
NM_001160176.3 | NP_001153648.1 | 4 | |
NM_001160179.2 | NP_001153651.1 | 5 | |
NM_001291962.1 | NP_001278891.1 | 6 | |
XM_006716410.3 | XP_006716473.1 | 3 | |
XM_011544687.1 | XP_011542989.1 | 6 | |
XM_011544688.1 | XP_011542990.1 | 5 | |
XM_011544689.2 | XP_011542991.1 | 2 | |
XM_017013947.1 | XP_016869436.1 | 8 |