gRNA Design Details

Target DNA Sequence
GAACCTTAACATCCATTGTG
PAM Sequence
GGG
Target locus
Chr.8: 18222161 - 18222183 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 86.93/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
NAT1
Entrez Gene ID
9
Gene Name
N-acetyltransferase 1
Gene Aliases
AAC1, MNAT, NAT-1, NATI
Chromosome Location
Chr.8: 18170419 - 18223689 on GRCh38
UniGene ID
Hs.591847
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000662.7 NP_000653.3 3
NM_001160170.3 NP_001153642.1 5
NM_001160171.3 NP_001153643.1 4
NM_001160172.3 NP_001153644.1 5
NM_001160173.3 NP_001153645.1 4
NM_001160174.2 NP_001153646.1 1
NM_001160175.3 NP_001153647.1 5
NM_001160176.3 NP_001153648.1 4
NM_001160179.2 NP_001153651.1 5
NM_001291962.1 NP_001278891.1 6
XM_006716410.3 XP_006716473.1 3
XM_011544687.1 XP_011542989.1 6
XM_011544688.1 XP_011542990.1 5
XM_011544689.2 XP_011542991.1 2
XM_017013947.1 XP_016869436.1 8