gRNA Design Details
- Target DNA Sequence
- GACATTTATCAGACTCTAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 75029685 - 75029707 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GCNT4
- Entrez Gene ID
-
51301
- Gene Name
- glucosaminyl (N-acetyl) transferase 4, core 2
- Gene Aliases
-
C2GNT3
- Chromosome Location
-
Chr.5: 75027464 - 75030899 on GRCh38
- UniGene ID
- Hs.272404
- Species
-
Human