gRNA Design Details
- Target DNA Sequence
- GGTTGTCTATAAATCAGCCC
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 186613208 - 186613230 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AHSG
- Entrez Gene ID
-
197
- Gene Name
- alpha 2-HS glycoprotein
- Gene Aliases
-
A2HS, AHS, FETUA, HSGA
- Chromosome Location
-
Chr.3: 186612928 - 186621318 on GRCh38
- UniGene ID
- Hs.324746
- Species
-
Human