gRNA Design Details
- Target DNA Sequence
- TTGATTGGTTCGTGCCTGAT
- PAM Sequence
- TGG
- Application
- Gene Knockout
Score:
89.03/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SATL1
- Entrez Gene ID
-
340562
- Gene Name
- spermidine/spermine N1-acetyl transferase-like 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 85092286 - 85116229 on GRCh38
- UniGene ID
- Hs.640783
- Species
-
Human