gRNA Design Details

Target DNA Sequence
GCAACGCCATTATGGGCAGT
PAM Sequence
GGG
Target locus
Chr.12: 46229203 - 46229225 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 91.05/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SLC38A1
Entrez Gene ID
81539
Gene Name
solute carrier family 38 member 1
Gene Aliases
ATA1, NAT2, SAT1, SNAT1
Chromosome Location
Chr.12: 46183055 - 46269425 on GRCh38
UniGene ID
Hs.533770
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001077484.1 NP_001070952.1 5
NM_001278387.1 NP_001265316.1 6
NM_001278388.1 NP_001265317.1 4
NM_001278389.1 NP_001265318.1 5
NM_001278390.1 NP_001265319.1 5
NM_030674.3 NP_109599.3 5
XM_011538784.2 XP_011537086.1 5
XM_011538786.2 XP_011537088.1 5
XM_011538787.2 XP_011537089.1 5
XM_017019990.1 XP_016875479.1 5