gRNA Design Details

Target DNA Sequence
TAATCACCTTCTCCGTTTTA
PAM Sequence
AGG
Target locus
Chr.9: 76502458 - 76502480 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 86.79/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GCNT1
Entrez Gene ID
2650
Gene Name
glucosaminyl (N-acetyl) transferase 1, core 2
Gene Aliases
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2
Chromosome Location
Chr.9: 76441666 - 76507416 on GRCh38
UniGene ID
Hs.521568
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001097633.1 NP_001091102.1 3
NM_001097634.1 NP_001091103.1 3
NM_001097635.1 NP_001091104.1 3
NM_001097636.1 NP_001091105.1 2
NM_001490.4 NP_001481.2 4
XM_017014620.1 XP_016870109.1 5
XM_017014621.1 XP_016870110.1 5