gRNA Design Details
- Target DNA Sequence
- AATGGTCGTGTTGCCAAAGT
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 119100679 - 119100701 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.29/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- DPAGT1
- Entrez Gene ID
-
1798
- Gene Name
- dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
- Gene Aliases
-
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT
- Chromosome Location
-
Chr.11: 119096503 - 119102414 on GRCh38
- UniGene ID
- Hs.524081
- Species
-
Human