gRNA Design Details

Target DNA Sequence
ACAGCACGGACCCTGAAAAT
PAM Sequence
GGG
Target locus
Chr.1: 165651027 - 165651049 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.77/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MGST3
Entrez Gene ID
4259
Gene Name
microsomal glutathione S-transferase 3
Gene Aliases
GST-III
Chromosome Location
Chr.1: 165630873 - 165656136 on GRCh38
UniGene ID
Hs.191734
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_004528.3 NP_004519.1 3
XM_005245174.3 XP_005245231.2 3