gRNA Design Details
- Target DNA Sequence
- AATCGCCCTCTTCTGCAGAC
- PAM Sequence
- AGG
- Target locus
-
Chr.20: 56497284 - 56497306 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.79/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GCNT7
- Entrez Gene ID
-
140687
- Gene Name
- glucosaminyl (N-acetyl) transferase family member 7
- Gene Aliases
-
C20orf105, dJ1153D9.2, gcnt
- Chromosome Location
-
Chr.20: 56491492 - 56525925 on GRCh38
- UniGene ID
- Hs.655916
- Species
-
Human