gRNA Design Details

Target DNA Sequence
AATCGCCCTCTTCTGCAGAC
PAM Sequence
AGG
Target locus
Chr.20: 56497284 - 56497306 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.79/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GCNT7
Entrez Gene ID
140687
Gene Name
glucosaminyl (N-acetyl) transferase family member 7
Gene Aliases
C20orf105, dJ1153D9.2, gcnt
Chromosome Location
Chr.20: 56491492 - 56525925 on GRCh38
UniGene ID
Hs.655916
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_080615.1 NP_542182.1 5