gRNA Design Details
- Target DNA Sequence
- GACCATGCGTTTTATCTGCT
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 41853497 - 41853519 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- OXCT1
- Entrez Gene ID
-
5019
- Gene Name
- 3-oxoacid CoA-transferase 1
- Gene Aliases
-
OXCT, SCOT
- Chromosome Location
-
Chr.5: 41730065 - 41870689 on GRCh38
- UniGene ID
- Hs.278277
- Species
-
Human