gRNA Design Details
- Target DNA Sequence
- AAGACCAACTGATGTAGTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 62402092 - 62402114 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- DIMT1
- Entrez Gene ID
-
27292
- Gene Name
- DIM1 dimethyladenosine transferase 1 homolog
- Gene Aliases
-
DIM1, DIMT1L, HSA9761, HUSSY5
- Chromosome Location
-
Chr.5: 62388523 - 62403939 on GRCh38
- UniGene ID
- Hs.731665
- Species
-
Human