gRNA Design Details
- Target DNA Sequence
- CTGACAGTAATGGATCTCAT
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 75789207 - 75789229 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
84.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RABGGTB
- Entrez Gene ID
-
5876
- Gene Name
- Rab geranylgeranyltransferase beta subunit
- Gene Aliases
-
GGTB
- Chromosome Location
-
Chr.1: 75786194 - 75795090 on GRCh38
- UniGene ID
- Hs.78948
- Species
-
Human