gRNA Design Details

Target DNA Sequence
GTCCATGACACTGGGGTACT
PAM Sequence
GGG
Target locus
Chr.1: 109656392 - 109656414 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 84.21/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GSTM4
Entrez Gene ID
2948
Gene Name
glutathione S-transferase mu 4
Gene Aliases
GSTM4-4, GTM4
Chromosome Location
Chr.1: 109656076 - 109665501 on GRCh38
UniGene ID
Hs.348387
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000850.4 NP_000841.1 1
NM_147148.2 NP_671489.1 1
NR_024538.1 1
XM_017001085.1 XP_016856574.1 1
XM_017001086.1 XP_016856575.1 1
XM_017001087.1 XP_016856576.1 1
XM_017001088.1 XP_016856577.1 1