gRNA Design Details
- Target DNA Sequence
- TTCAAGTGGGCATCGAGGCT
- PAM Sequence
- GGG
- Target locus
-
Chr.19: 8955389 - 8955411 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tut1
- Entrez Gene ID
-
70044
- Gene Name
- terminal uridylyl transferase 1, U6 snRNA-specific
- Gene Aliases
-
2700038E08Rik, PAPD2, Rbm21, TUTase6
- Chromosome Location
-
Chr.19: 8953850 - 8966210 on GRCm38
- UniGene ID
- Mm.165979
- Species
-
Mouse