gRNA Design Details
- Target DNA Sequence
- TTTTCCTCAGGTCGACATAA
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 52899998 - 52900020 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
81.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTA3
- Entrez Gene ID
-
2940
- Gene Name
- glutathione S-transferase alpha 3
- Gene Aliases
-
GSTA3-3, GTA3
- Chromosome Location
-
Chr.6: 52896639 - 52909799 on GRCh38
- UniGene ID
- Hs.102484
- Species
-
Human