gRNA Design Details
- Target DNA Sequence
- GATCGCCAACAACACCTTCA
- PAM Sequence
- CGG
- Target locus
-
Chr.16: 1362056 - 1362078 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
93.4/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GNPTG
- Entrez Gene ID
-
84572
- Gene Name
- N-acetylglucosamine-1-phosphate transferase gamma subunit
- Gene Aliases
-
C16orf27, GNPTAG, LP2537, RJD9
- Chromosome Location
-
Chr.16: 1351899 - 1363351 on GRCh38
- UniGene ID
- Hs.241575
- Species
-
Human