gRNA Design Details
- Target DNA Sequence
- GTCAGAAATCATACGCACGC
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 77327985 - 77328007 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTZ1
- Entrez Gene ID
-
2954
- Gene Name
- glutathione S-transferase zeta 1
- Gene Aliases
-
GSTZ1-1, MAAI, MAI
- Chromosome Location
-
Chr.14: 77320887 - 77331597 on GRCh38
- UniGene ID
- Hs.655292
- Species
-
Human