gRNA Design Details

Target DNA Sequence
GGCACTGAATCCTATGAAGC
PAM Sequence
AGG
Target locus
Chr.14: 77327489 - 77327511 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 89.5/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GSTZ1
Entrez Gene ID
2954
Gene Name
glutathione S-transferase zeta 1
Gene Aliases
GSTZ1-1, MAAI, MAI
Chromosome Location
Chr.14: 77320887 - 77331597 on GRCh38
UniGene ID
Hs.655292
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001312660.1 NP_001299589.1 5
NM_145870.2 NP_665877.1 4
NM_145871.2 NP_665878.2 4
XM_011536670.2 XP_011534972.1 5
XM_011536671.2 XP_011534973.1 4
XM_017021243.1 XP_016876732.1 4