gRNA Design Details
- Target DNA Sequence
- GTACGGCAGCTGGAGAGAAC
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 535167 - 535189 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.15/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- B4GALNT3
- Entrez Gene ID
-
283358
- Gene Name
- beta-1,4-N-acetyl-galactosaminyltransferase 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 460377 - 561892 on GRCh38
- UniGene ID
- Hs.504416
- Species
-
Human