gRNA Design Details
- Target DNA Sequence
- GATGGTCTACCCCCAGCCAA
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 133258110 - 133258132 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.78/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ABO
- Entrez Gene ID
-
28
- Gene Name
- ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
- Gene Aliases
-
A3GALNT, A3GALT1, GTB, NAGAT
- Chromosome Location
-
Chr.9: 133255176 - 133275214 on GRCh38
- UniGene ID
- Hs.654423
- Species
-
Human