gRNA Design Details
- Target DNA Sequence
- TTGCTTGAGTTGTAACAAAT
- PAM Sequence
- CGG
- Target locus
-
Chr.18: 12371664 - 12371686 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
81.31/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AFG3L2
- Entrez Gene ID
-
10939
- Gene Name
- AFG3 like matrix AAA peptidase subunit 2
- Gene Aliases
-
SCA28, SPAX5
- Chromosome Location
-
Chr.18: 12328944 - 12377309 on GRCh38
- UniGene ID
- Hs.726355
- Species
-
Human