gRNA Design Details
- Target DNA Sequence
- AGTGAACCTAAAGAAGTTAT
- PAM Sequence
- GGG
- Target locus
-
Chr.18: 12370860 - 12370882 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
73.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AFG3L2
- Entrez Gene ID
-
10939
- Gene Name
- AFG3 like matrix AAA peptidase subunit 2
- Gene Aliases
-
SCA28, SPAX5
- Chromosome Location
-
Chr.18: 12328944 - 12377309 on GRCh38
- UniGene ID
- Hs.726355
- Species
-
Human