gRNA Design Details
- Target DNA Sequence
- ACGTACCCACAGCACTATTA
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 24942375 - 24942397 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.3/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRTFDC1
- Entrez Gene ID
-
56952
- Gene Name
- phosphoribosyl transferase domain containing 1
- Gene Aliases
-
HHGP
- Chromosome Location
-
Chr.10: 24848607 - 24952644 on GRCh38
- UniGene ID
- Hs.405619
- Species
-
Human