gRNA Design Details
- Target DNA Sequence
- GGCCATCGCCTACTCATTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 991424 - 991446 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.43/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SLC26A1
- Entrez Gene ID
-
10861
- Gene Name
- solute carrier family 26 member 1
- Gene Aliases
-
CAON, EDM4, SAT-1, SAT1
- Chromosome Location
-
Chr.4: 979073 - 993440 on GRCh38
- UniGene ID
- Hs.658244
- Species
-
Human