gRNA Design Details
- Target DNA Sequence
- TCTTTTCGAGGCGGAGGTCG
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 109816168 - 109816190 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GAR1
- Entrez Gene ID
-
54433
- Gene Name
- GAR1 ribonucleoprotein
- Gene Aliases
-
NOLA1
- Chromosome Location
-
Chr.4: 109815510 - 109824737 on GRCh38
- UniGene ID
- Hs.69851
- Species
-
Human