gRNA Design Details
- Target DNA Sequence
- GGTCACGAAGCCAAAGCAAC
- PAM Sequence
- GGG
- Target locus
-
Chr.5: 137753905 - 137753927 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HNRNPA0
- Entrez Gene ID
-
10949
- Gene Name
- heterogeneous nuclear ribonucleoprotein A0
- Gene Aliases
-
HNRPA0
- Chromosome Location
-
Chr.5: 137751384 - 137754350 on GRCh38
- UniGene ID
- Hs.645902
- Species
-
Human