gRNA Design Details

Target DNA Sequence
AACTCCATCACCAGCAGGTA
PAM Sequence
GGG
Target locus
Chr.17: 81127593 - 81127615 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.23/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AATK
Entrez Gene ID
9625
Gene Name
apoptosis associated tyrosine kinase
Gene Aliases
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP
Chromosome Location
Chr.17: 81117295 - 81166072 on GRCh38
UniGene ID
Hs.514575
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001080395.2 NP_001073864.2 6
NM_004920.2 NP_004911.2 5
XM_006722193.2 XP_006722256.1 6
XM_006722194.2 XP_006722257.1 6
XM_006722195.2 XP_006722258.1 4
XM_006722196.2 XP_006722259.1 6
XM_011525509.2 XP_011523811.1 5
XM_011525510.2 XP_011523812.1 5
XM_017025421.1 XP_016880910.1 5
XM_017025422.1 XP_016880911.1 4