gRNA Design Details

Target DNA Sequence
GCCTGGTAGCGTTGAGATGT
PAM Sequence
GGG
Target locus
Chr.10: 104279392 - 104279414 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.06/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GSTO2
Entrez Gene ID
119391
Gene Name
glutathione S-transferase omega 2
Gene Aliases
GSTO 2-2, bA127L20.1
Chromosome Location
Chr.10: 104267626 - 104304948 on GRCh38
UniGene ID
Hs.107384
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001191014.1 NP_001177943.1 3
NM_183239.1 NP_899062.1 5
XM_006717624.3 XP_006717687.1 5
XM_011539270.2 XP_011537572.1 5
XM_011539271.1 XP_011537573.1 5
XM_011539272.2 XP_011537574.1 5