gRNA Design Details
- Target DNA Sequence
- CGTTGCAGGAGCATTGGCTA
- PAM Sequence
- CGG
- Target locus
-
Chr.7: 100159343 - 100159365 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
82.56/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Lipt2
- Entrez Gene ID
-
67164
- Gene Name
- lipoyl(octanoyl) transferase 2 (putative)
- Gene Aliases
-
2610209A20Rik
- Chromosome Location
-
Chr.7: 100159277 - 100160931 on GRCm38
- UniGene ID
- Mm.46777
- Species
-
Mouse