gRNA Design Details
- Target DNA Sequence
- TCCGGAGTCCAGTGCTCTTT
- PAM Sequence
- CGG
- Application
- Gene Knockout
Score:
89.32/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SAT1
- Entrez Gene ID
-
6303
- Gene Name
- spermidine/spermine N1-acetyltransferase 1
- Gene Aliases
-
DC21, KFSD, KFSDX, SAT, SSAT, SSAT-1
- Chromosome Location
-
Chr.X: 23783158 - 23786223 on GRCh38
- UniGene ID
- Hs.28491
- Species
-
Human