gRNA Design Details

Target DNA Sequence
TCTGATCGTTCTACTACTGC
PAM Sequence
AGG
Target locus
Chr.4: 105717884 - 105717906 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 88.49/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GSTCD
Entrez Gene ID
79807
Gene Name
glutathione S-transferase C-terminal domain containing
Gene Aliases
-
Chromosome Location
Chr.4: 105708778 - 105848023 on GRCh38
UniGene ID
Hs.161429
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001031720.3 NP_001026890.2 2
XM_005263222.3 XP_005263279.1 2
XM_011532248.2 XP_011530550.1 2
XM_011532249.2 XP_011530551.1 2
XM_011532252.2 XP_011530554.1 2