gRNA Design Details

Target DNA Sequence
TATACAGGTTCCACCAAAGA
PAM Sequence
AGG
Target locus
Chr.10: 104297600 - 104297622 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.81/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GSTO2
Entrez Gene ID
119391
Gene Name
glutathione S-transferase omega 2
Gene Aliases
GSTO 2-2, bA127L20.1
Chromosome Location
Chr.10: 104267626 - 104304948 on GRCh38
UniGene ID
Hs.107384
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001191013.1 NP_001177942.1 5
NM_001191014.1 NP_001177943.1 4
NM_001191015.1 NP_001177944.1 3
NM_183239.1 NP_899062.1 6
XM_011539270.2 XP_011537572.1 6