gRNA Design Details
- Target DNA Sequence
- TCTTGCTCGAGATGTGATGA
- PAM Sequence
- AGG
- Application
- Gene Knockout
Score:
87.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HPRT1
- Entrez Gene ID
-
3251
- Gene Name
- hypoxanthine phosphoribosyltransferase 1
- Gene Aliases
-
HGPRT, HPRT
- Chromosome Location
-
Chr.X: 134460145 - 134500668 on GRCh38
- UniGene ID
- Hs.412707
- Species
-
Human