gRNA Design Details
- Target DNA Sequence
- ATTTCCGCACACAACTGATA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 109738112 - 109738134 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.53/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTM3
- Entrez Gene ID
-
2947
- Gene Name
- glutathione S-transferase mu 3
- Gene Aliases
-
GST5, GSTB, GSTM3-3, GTM3
- Chromosome Location
-
Chr.1: 109733932 - 109741038 on GRCh38
- UniGene ID
- Hs.2006
- Species
-
Human