gRNA Design Details
- Target DNA Sequence
- GTCATTCACCATGTCCACCA
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 67585168 - 67585190 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.57/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTP1
- Entrez Gene ID
-
2950
- Gene Name
- glutathione S-transferase pi 1
- Gene Aliases
-
DFN7, FAEES3, GST3, GSTP, HEL-S-22, PI
- Chromosome Location
-
Chr.11: 67583595 - 67586653 on GRCh38
- UniGene ID
- Hs.523836
- Species
-
Human