gRNA Design Details

Target DNA Sequence
AGAACCATCTTCCCTCTGGG
PAM Sequence
GGG
Target locus
Chr.22: 46350371 - 46350393 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 85.65/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TRMU
Entrez Gene ID
55687
Gene Name
tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Gene Aliases
LCAL3, MTO2, MTU1, TRMT, TRMT1, TRNT1
Chromosome Location
Chr.22: 46335401 - 46357340 on GRCh38
UniGene ID
Hs.439524
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001282782.1 NP_001269711.1 4
NM_001282783.1 NP_001269712.1 4
NM_001282784.1 NP_001269713.1 4
NM_001282785.1 NP_001269714.1 5
NM_018006.4 NP_060476.2 5
NR_104240.1 5
NR_104241.1 4
XM_005261678.1 XP_005261735.1 5
XM_005261681.1 XP_005261738.1 5
XM_011530271.2 XP_011528573.1 5
XM_011530272.1 XP_011528574.1 5
XM_011530273.1 XP_011528575.1 5
XM_011530274.2 XP_011528576.1 4
XM_011530275.1 XP_011528577.1 5