gRNA Design Details
- Target DNA Sequence
- TTTTCCTCAGGTGGACATAC
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 52794189 - 52794211 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
76.39/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTA1
- Entrez Gene ID
-
2938
- Gene Name
- glutathione S-transferase alpha 1
- Gene Aliases
-
GST-epsilon, GST2, GSTA1-1, GTH1
- Chromosome Location
-
Chr.6: 52791380 - 52803967 on GRCh38
- UniGene ID
- Hs.446309
- Species
-
Human