gRNA Design Details
- Target DNA Sequence
- ACTGAAACCAGAATACCTGC
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 109671292 - 109671314 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
79.52/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTM2
- Entrez Gene ID
-
2946
- Gene Name
- glutathione S-transferase mu 2 (muscle)
- Gene Aliases
-
GST4, GSTM, GSTM2-2, GTHMUS
- Chromosome Location
-
Chr.1: 109668022 - 109683997 on GRCh38
- UniGene ID
- Hs.279837
- Species
-
Human