gRNA Design Details

Target DNA Sequence
ACTACTGAAGGAACTACAGC
PAM Sequence
AGG
Target locus
Chr.12: 101789975 - 101789997 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 85.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GNPTAB
Entrez Gene ID
79158
Gene Name
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
Gene Aliases
GNPTA, ICD
Chromosome Location
Chr.12: 101745497 - 101830867 on GRCh38
UniGene ID
Hs.46850
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_024312.4 NP_077288.2 3
XM_006719593.2 XP_006719656.1 3
XM_011538731.2 XP_011537033.1 3
XM_017019961.1 XP_016875450.1 3
XM_017019962.1 XP_016875451.1 3