Gene Transcripts

Gene Symbol
LOC501038
Entrez Gene ID
501038
Gene Name
Ab2-060
Gene Aliases
Chromosome Location
Chr. 8: 103341326 - 103351707 on Build Rnor_6.0
UniGene ID
Rn.43844
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047965.1 NP_001041430.1 2 67
XM_006243552.2 XP_006243614.1 2 1594
GenBank mRNA AY325184.1 - 2 67

View Full Product Details