Gene Transcripts

Gene Symbol
Ip6k1
Entrez Gene ID
50560
Gene Name
inositol hexakisphosphate kinase 1
Gene Aliases
Ihpk1, Itpk6
Chromosome Location
Chr. 8: 116781957 - 116826152 on Build Rnor_6.0
UniGene ID
Rn.16751
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053316.2 NP_445768.1 2 265
XM_006243839.1 XP_006243901.1 2 467
GenBank mRNA AB049151.1 - 2 316
BC078702.1 - 2 265 7132706

View Full Product Details