Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_004712.4 | NP_004703.1 |
XM_011525463.2 | XP_011523765.1 | |
XM_017025297.1 | XP_016880786.1 | |
XM_017025298.1 | XP_016880787.1 | |
GenBank mRNA | AF260566.1 | - |
AI365104.1 | - | |
AK021483.1 | - | |
AK097197.1 | - | |
AK294190.1 | - | |
AK303800.1 | - | |
BC003565.1 | - | |
BT009754.1 | - | |
D84064.1 | - | |
DR001213.1 | - | |
ENST00000329138.0 | - | |
KU178466.1 | - | |
KU178467.1 | - | |
U43895.1 | - |