Gene Transcripts

Gene Symbol
PGF
Entrez Gene ID
5228
Gene Name
placental growth factor
Gene Aliases
D12S1900, PGFL, PIGF, PLGF, PlGF-2, SHGC-10760
Chromosome Location
Chr.14: 74941830 - 74955764 on Build GRCh38
Uni Gene
Hs.252820
Interrogated Sequence Translated Protein
RefSeq NM_001207012.1 NP_001193941.1
NM_001293643.1 NP_001280572.1
NM_002632.5 NP_002623.2
GenBank mRNA A18411.1 -
AK023843.1 -
AK309886.1 -
AK310608.1 -
AK310886.1 -
AL543693.3 -
BC001422.2 -
BC007255.1 -
BC007789.2 -
BT007182.1 -
BX248289.1 -
BX356823.2 -
ENST00000555567.0 -
S72960.1 -
X54936.1 -

View Full Product Details