Gene Transcripts

Gene Symbol
NGF
Entrez Gene ID
4803
Gene Name
nerve growth factor
Gene Aliases
Beta-NGF, HSAN5, NGFB
Chromosome Location
Chr.1: 115285915 - 115338253 on Build GRCh38
Uni Gene
Hs.2561
Interrogated Sequence Translated Protein
RefSeq NM_002506.2 NP_002497.2
XM_006710663.3 XP_006710726.1
XM_011541518.2 XP_011539820.1
GenBank mRNA AF411526.1 -
BC032517.2 -
BC126148.1 -
BC126150.1 -
BE045524.1 -
BT019733.1 -
CR541855.1 -
ENST00000369512.0 -
X52599.1 -

View Full Product Details