Gene Transcripts

Gene Symbol
HGS
Entrez Gene ID
9146
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
Gene Aliases
HRS
Chromosome Location
Chr.17: 81683932 - 81702121 on Build GRCh38
Uni Gene
Hs.661056
Interrogated Sequence Translated Protein
RefSeq NM_004712.4 NP_004703.1
XM_011525463.2 XP_011523765.1
XM_017025297.1 XP_016880786.1
XM_017025298.1 XP_016880787.1
GenBank mRNA AF260566.1 -
AI365104.1 -
AK021483.1 -
AK097197.1 -
AK294190.1 -
AK303800.1 -
BC003565.1 -
BT009754.1 -
D84064.1 -
DR001213.1 -
ENST00000329138.0 -
KU178466.1 -
KU178467.1 -
U43895.1 -

View Full Product Details